Myosmine, 98%, may contain up to 2% water
Myosmine, 98%, may contain up to 2% water
Myosmine, 98%, may contain up to 2% water
Thermo Scientific Chemicals

Myosmine, 98%, may contain up to 2% water

CAS: 532-12-7 | C9H10N2 | 146.193 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number L11886.MC
also known as L11886-MC
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.00
Save 12.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
73.65
Special offer
Online exclusive
Ends: 28-Dec-2026
86.00
Save 12.35
Each
Chemical Identifiers
CAS532-12-7
IUPAC Name3-(3,4-dihydro-2H-pyrrol-5-yl)pyridine
Molecular FormulaC9H10N2
InChI KeyDPNGWXJMIILTBS-UHFFFAOYSA-N
SMILESC1CN=C(C1)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream or pale yellow to yellow or orange to brown
FormPowder or crystalline powder
Assay (GC)≥97.5%
Water Content (Karl Fischer Titration)<2.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only