2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol
2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol
2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol
Thermo Scientific Chemicals

2-Methoxyethyl acrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol

CAS: 3121-61-7 | C6H10O3 | 130.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number L11902.18
also known as L11902-18
Price (USD)
21.40
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
21.40
Each
Chemical Identifiers
CAS3121-61-7
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Solution Test10% soln. in Diethyl ether: Clear
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4260-1.4300 @ 20?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only