2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, 98%, stab.
2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, 98%, stab.
2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, 98%, stab.
2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, 98%, stab.
Thermo Scientific Chemicals

2,2,3,3,4,4,5,5-Octafluoropentyl methacrylate, 98%, stab.

CAS: 355-93-1 | C9H8F8O2 | 300.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L12044.06
also known as L12044-06
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.70
Save 13.05 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.70
Save 13.05 (15%)
Each
Add to cart
Chemical Identifiers
CAS355-93-1
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3565-1.3610 @ 20?C
Solution TestCLEAR: 10% in Methanol
Assay (GC)≥97.5%
Stabilizer4-Methoxyphenol or BHT
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only