3-Chloro-4-methylphenyl isocyanate, 98%
3-Chloro-4-methylphenyl isocyanate, 98%
3-Chloro-4-methylphenyl isocyanate, 98%
Thermo Scientific Chemicals

3-Chloro-4-methylphenyl isocyanate, 98%

CAS: 28479-22-3 | C8H6ClNO | 167.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L12064.06
also known as L12064-06
Price (USD)
49.65
Online Exclusive
54.50
Save 4.85 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.50
Save 4.85 (9%)
Each
Chemical Identifiers
CAS28479-22-3
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5540-1.5580?C (as melt)
Assay (GC)>97.5%
FormLiquid (low melting solid)
Appearance (Color)Clear or slightly turbid, colorless to yellow
CommentMay form small amount of sediment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only