5-Phenyl-1-pentanol, 97%
5-Phenyl-1-pentanol, 97%
5-Phenyl-1-pentanol, 97%
Thermo Scientific Chemicals

5-Phenyl-1-pentanol, 97%

CAS: 10521-91-2 | C11H16O | 164.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12070.06
also known as L12070-06
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
98.65
Online Exclusive
109.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS10521-91-2
IUPAC Name5-phenylpentan-1-ol
Molecular FormulaC11H16O
InChI KeyDPZMVZIQRMVBBW-UHFFFAOYSA-N
SMILESOCCCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay (GC)≥96.0%
Refractive Index1.5110-1.5170 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
RUO – Research Use Only
Hidetaka Nagatomo; Yoh-ichi Matsushita; Kazuhiro Sugamoto; Takanao Matsui. Preparation and properties of gelatin-immobilized beta-glucosidase from Pyrococcus furiosus. Bioscience, Biotechnology, and Biochemistry.2005, 69, (1), 128-136.