2-n-Hexyldecanoic acid, 97%
2-n-Hexyldecanoic acid, 97%
2-n-Hexyldecanoic acid, 97%
Thermo Scientific Chemicals

2-n-Hexyldecanoic acid, 97%

CAS: 25354-97-6 | C16H32O2 | 256.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L12238.14
also known as L12238-14
Price (USD)
31.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
31.60
Each
Chemical Identifiers
CAS25354-97-6
IUPAC Name2-hexyldecanoic acid
Molecular FormulaC16H32O2
InChI KeyJMOLZNNXZPAGBH-UHFFFAOYNA-N
SMILESCCCCCCCCC(CCCCCC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid or fused solid
Assay (Silylated GC)≥96.0% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Refractive Index1.4435-1.4485 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only