N-[4-(Trifluoromethyl)phenyl]thiourea, 98%
N-[4-(Trifluoromethyl)phenyl]thiourea, 98%
N-[4-(Trifluoromethyl)phenyl]thiourea, 98%
Thermo Scientific Chemicals

N-[4-(Trifluoromethyl)phenyl]thiourea, 98%

CAS: 1736-72-7 | C8H7F3N2S | 220.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12355.06
also known as L12355-06
Price (USD)
132.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
132.00
Each
Specifications
Beilstein2695479
CAS1736-72-7
Chemical Name or MaterialN-[4-(Trifluoromethyl)phenyl]thiourea
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Formula Weight220.22
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only