N-(4-Hydroxyphenyl)thiourea, 98+%
N-(4-Hydroxyphenyl)thiourea, 98+%
N-(4-Hydroxyphenyl)thiourea, 98+%
N-(4-Hydroxyphenyl)thiourea, 98+%
Thermo Scientific Chemicals

N-(4-Hydroxyphenyl)thiourea, 98+%

CAS: 1520-27-0 | C7H8N2OS | 168.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12537.06
also known as L12537-06
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1520-27-0
IUPAC Name(4-hydroxyphenyl)thiourea
Molecular FormulaC7H8N2OS
InChI KeyQICKOOCQSYZYQB-UHFFFAOYSA-N
SMILESNC(=S)NC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay (HPLC)≥98.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only