1H-Indene, 97%
1H-Indene, 97%
1H-Indene, 97%
Thermo Scientific Chemicals

1H-Indene, 97%

CAS: 95-13-6 | C9H8 | 116.163 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L12665.14
also known as L12665-14
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
152.65
Online Exclusive
169.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS95-13-6
IUPAC Name1H-indene
Molecular FormulaC9H8
InChI KeyYBYIRNPNPLQARY-UHFFFAOYSA-N
SMILESC1C=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>96.0%
Refractive Index1.5720-1.5770 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to yellow
1H-Indene is used in the synthesis of new C60 derivative, indene-C60 bisadduct1. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2. It is also used in the synthesis of istatins and in the production of indene/cumarone thermoplastic resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Indene is used in the synthesis of new C60 derivative, indene-C60 bisadduct1. It was used in preparing polyindene by the controlled cationic polymerization initiated with cumyl methyl ether/TiCl4 in CH2Cl2. It is also used in the synthesis of istatins and in the production of indene/cumarone thermoplastic resins.

Solubility
Soluble in ethanol, acetone, ethyl ether, and ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which areopened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents Recommended storage temperature is 2 - 8°C. It is sensitive to light.
RUO – Research Use Only

General References:

  1. Hahn SF.; Hillmyer MA. High glass transition temperature polyolefins obtained by the catalytic hydrogenation of polyindene. Macromolecules. 2003, 36 (1), 71-76.
  2. Youjun He.; Hsiang-Yu Chen.; Jianhui Hou.; Yongfang Li. Indene-C(60) bisadduct: a new acceptor for high-performance polymer solar cells. Journal of the American Chemical Society. 2010, 132 (4), 1377-1382.