4-Bromo-2-ethylaniline, 97%
4-Bromo-2-ethylaniline, 97%
4-Bromo-2-ethylaniline, 97%
4-Bromo-2-ethylaniline, 97%
Thermo Scientific Chemicals

4-Bromo-2-ethylaniline, 97%

CAS: 45762-41-2 | C8H10BrN | 200.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L12725.14
also known as L12725-14
Price (USD)
134.65
Special offer
Online exclusive
Ends: 30-Jun-2026
158.00
Save 23.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
134.65
Special offer
Online exclusive
Ends: 30-Jun-2026
158.00
Save 23.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS45762-41-2
IUPAC Name4-bromo-2-ethylaniline
Molecular FormulaC8H10BrN
InChI KeyLGOZNQPHTIGMQJ-UHFFFAOYSA-N
SMILESCCC1=C(N)C=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5975-1.6025 @ 20?C
Appearance (Color)Clear colorless to yellow to dark brown
Assay (GC)≥96.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only