N,N-Di-n-butylformamide, 99%
N,N-Di-n-butylformamide, 99%
N,N-Di-n-butylformamide, 99%
N,N-Di-n-butylformamide, 99%
Thermo Scientific Chemicals

N,N-Di-n-butylformamide, 99%

CAS: 761-65-9 | C9H19NO | 157.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L12755.14
also known as L12755-14
Price (USD)
33.65
Online Exclusive
37.60
Save 3.95 (11%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.60
Save 3.95 (11%)
Each
Add to cart
Chemical Identifiers
CAS761-65-9
IUPAC NameN,N-dibutylformamide
Molecular FormulaC9H19NO
InChI KeyNZMAJUHVSZBJHL-UHFFFAOYSA-N
SMILESCCCCN(CCCC)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4425-1.4460 @ 20?C
FormLiquid
Identification (FTIR)Conforms
N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. George C.Levy; Gordon L.Nelson. Carbon-13 NMR study of aliphatic amides and oximes. Spin-lattice relaxation times and fast internal motions. J. Am. Chem. Soc. 1972, 94,(14), 4897-4901.
  2. Nhat Quang Nguyen-Trung; Oliver Botta; Silvia Terenzi; Peter Strazewski. A practical route to 3‘-amino-3‘-deoxyadenosine derivatives and puromycin analogues. J. Org. Chem. 2003, 68,(5), 2038-2041.