2-Amino-4-hydroxyquinoline hydrate, 97%, water <12%
2-Amino-4-hydroxyquinoline hydrate, 97%, water <12%
2-Amino-4-hydroxyquinoline hydrate, 97%, water <12%
Thermo Scientific Chemicals

2-Amino-4-hydroxyquinoline hydrate, 97%, water <12%

CAS: 42712-64-1 | C9H8N2O | 160.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12760.03
also known as L12760-03
Price (USD)
26.65
Online Exclusive
29.40
Save 2.75 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
26.65
Online Exclusive
29.40
Save 2.75 (9%)
Each
Chemical Identifiers
CAS42712-64-1
IUPAC Name2-amino-1,4-dihydroquinolin-4-one
Molecular FormulaC9H8N2O
InChI KeyLWGUCIXHBVVATR-UHFFFAOYSA-N
SMILESNC1=CC(=O)C2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤12%
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0% (dry wt. basis)
Appearance (Color)White to pale brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only