2,6-Dimethylphenyl isocyanate, 98%
2,6-Dimethylphenyl isocyanate, 98%
2,6-Dimethylphenyl isocyanate, 98%
Thermo Scientific Chemicals

2,6-Dimethylphenyl isocyanate, 98%

CAS: 28556-81-2 | C9H9NO | 147.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L12805.06
also known as L12805-06
Price (USD)
72.65
Online Exclusive
80.50
Save 7.85 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.50
Save 7.85 (10%)
Each
Chemical Identifiers
CAS28556-81-2
IUPAC Name2-isocyanato-1,3-dimethylbenzene
Molecular FormulaC9H9NO
InChI KeyYQLRKXVEALTVCZ-UHFFFAOYSA-N
SMILESCC1=CC=CC(C)=C1N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5345-1.5385 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only