Allyl ethyl sulfide, 97%
Allyl ethyl sulfide, 97%
Allyl ethyl sulfide, 97%
Thermo Scientific Chemicals

Allyl ethyl sulfide, 97%

CAS: 5296-62-8 | C5H10S | 102.20 g/mol
Have Questions?
Catalog NumberQuantity
L12847.06
also known as L12847-06
5 g
Catalog number L12847.06
also known as L12847-06
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 30-Jun-2026
104.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS5296-62-8
IUPAC Name3-(ethylsulfanyl)prop-1-ene
Molecular FormulaC5H10S
InChI KeyNOJXPGXFDASWEI-UHFFFAOYSA-N
SMILESCCSCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4655-1.4705 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only