5-Hexenoic acid, 99%
5-Hexenoic acid, 99%
5-Hexenoic acid, 99%
5-Hexenoic acid, 99%
Thermo Scientific Chemicals

5-Hexenoic acid, 99%

CAS: 1577-22-6 | C6H10O2 | 114.144 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12863.06
also known as L12863-06
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
175.65
Online Exclusive
195.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1577-22-6
IUPAC Namehex-5-enoic acid
Molecular FormulaC6H10O2
InChI KeyXUDOZULIAWNMIU-UHFFFAOYSA-N
SMILESOC(=O)CCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4335-1.4365 @ 20?C
5-Hexenoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hexenoic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Olivier Persenaire; Michael Alexandre; Philippe Degee; Philippe Dubois. Mechanisms and kinetics of thermal degradation of poly(ε-caprolactone). Biomacromolecules. 2001, 2,(1), 288-294.