4-n-Butoxyphenyl isocyanate, 98%
4-n-Butoxyphenyl isocyanate, 98%
4-n-Butoxyphenyl isocyanate, 98%
4-n-Butoxyphenyl isocyanate, 98%
Thermo Scientific Chemicals

4-n-Butoxyphenyl isocyanate, 98%

CAS: 28439-86-3 | C11H13NO2 | 191.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12877.06
also known as L12877-06
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS28439-86-3
IUPAC Name1-butoxy-4-isocyanatobenzene
Molecular FormulaC11H13NO2
InChI KeyASFIDVOMDQBCNP-UHFFFAOYSA-N
SMILESCCCCOC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to yellow
FormLiquid
Refractive Index1.5200-1.5240 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only