2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.
Thermo Scientific Chemicals

2,2,3,3,4,4,4-Heptafluorobutyl methacrylate, 97%, stab.

CAS: 13695-31-3 | C8H7F7O2 | 268.131 g/mol
Have Questions?
Catalog NumberQuantity
L12922.1425 g
Catalog number L12922.14
Price (USD)
342.65
Online Exclusive
380.00
Save 37.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
342.65
Online Exclusive
380.00
Save 37.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS13695-31-3
IUPAC Name2,2,3,3,4,4,4-heptafluorobutyl 2-methylprop-2-enoate
Molecular FormulaC8H7F7O2
InChI KeyVIEHKBXCWMMOOU-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OCC(F)(F)C(F)(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Solution Test10% in Diethyl ether: Clear
Solution Test10% in Methanol: Clear
Refractive Index1.3395-1.3445 @ 20?C
View more
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,3,3,4,4,4-Heptafluorobutyl methacrylate is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. Zhenghong Luo; Tengyun He; Haijiang Yu; Lizong Dai. A novel ABC triblock copolymer with very low surface energy: Poly(dimethylsiloxane)-block-Poly(methyl methacrylate)-block-Poly(2,2,3,3,4,4,4-heptafluorobutyl methacrylate). Macromolecular Reaction Engineering. 2008, 2,(5), 398-406.
  2. A.Benedetto; P.Viel; S.Noel; N.Izard; P.Chenevier; S.Palacin. Carbon nanotubes/fluorinated polymers nanocomposite thin films for electrical contacts lubrication. Surface Science. 2007, 601,(18), 3687-3692.