Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L12925.14
also known as L12925-14
25 g
Catalog number L12925.14
also known as L12925-14
Price (USD)
82.65
Online Exclusive
92.20
Save 9.55 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
82.65
Online Exclusive
92.20
Save 9.55 (10%)
Each
Specifications
Beilstein1937528
CAS3066-71-5
Chemical Name or MaterialCyclohexyl acrylate
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Density0.98
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only