(Propargyloxy)trimethylsilane, 97%
(Propargyloxy)trimethylsilane, 97%
(Propargyloxy)trimethylsilane, 97%
Thermo Scientific Chemicals

(Propargyloxy)trimethylsilane, 97%

CAS: 5582-62-7 | C6H12OSi | 128.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L12936.06
also known as L12936-06
Price (USD)
41.65
Online Exclusive
46.30
Save 4.65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
46.30
Save 4.65 (10%)
Each
Chemical Identifiers
CAS5582-62-7
IUPAC Nametrimethyl(prop-2-yn-1-yloxy)silane
Molecular FormulaC6H12OSi
InChI KeyZZRPJWCNCLSOLR-UHFFFAOYSA-N
SMILESC[Si](C)(C)OCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4060-1.4110 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to yellow to red
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only