4-Phenyl-1-butyne, 98%
4-Phenyl-1-butyne, 98%
4-Phenyl-1-butyne, 98%
Thermo Scientific Chemicals

4-Phenyl-1-butyne, 98%

CAS: 16520-62-0 | C10H10 | 130.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L12946.06
also known as L12946-06
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
92.65
Special offer
Online exclusive
Ends: 30-Jun-2026
109.00
Save 16.35 (15%)
Each
Chemical Identifiers
CAS16520-62-0
IUPAC Name(but-3-yn-1-yl)benzene
Molecular FormulaC10H10
InChI KeyQDEOKXOYHYUKMS-UHFFFAOYSA-N
SMILESC#CCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5200-1.5240 @ 20°C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥96.0%
FormLiquid
4-Phenyl-1-butyne is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-butyne is used as an intermediate in chemical research.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Keith D. King.; Tam T. Nguyen. Very low-pressure pyrolysis of 4-phenyl-1-butyne. The propargyl resonance energy. J. Phys. Chem. 1979, 83 (15), 1940-1943.
  2. Se-Young Oh.; Fumiteru Oguri.; Kazuo Akagi.; Hideki Shirakawa. Polymerization of 4-phenyl-1-butyne catalyzed by metathesis and Ziegler-Natta catalysts. Journal of Polymer Science Part A: Polymer Chemistry. 1991, 31 (3), 781-787.