2-Chloro-4-fluorobenzylamine, 97%
2-Chloro-4-fluorobenzylamine, 97%
2-Chloro-4-fluorobenzylamine, 97%
Thermo Scientific Chemicals

2-Chloro-4-fluorobenzylamine, 97%

CAS: 15205-11-5 | C7H7ClFN | 159.588 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L12957.06
also known as L12957-06
Price (USD)
251.65
Special offer
Online exclusive
Ends: 30-Jun-2026
296.00
Save 44.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
251.65
Special offer
Online exclusive
Ends: 30-Jun-2026
296.00
Save 44.35 (15%)
Each
Chemical Identifiers
CAS15205-11-5
IUPAC Name1-(2-chloro-4-fluorophenyl)methanamine
Molecular FormulaC7H7ClFN
InChI KeyCBKWAXKMZUULLO-UHFFFAOYSA-N
SMILESNCC1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
FormLiquid
CommentMaterial sourced in the U.S. and in other countries
Refractive Index1.5320-1.5370 @ 20°C (non-U.S. sourced material)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only