3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane, 97%, Thermo Scientific Chemicals
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane, 97%, Thermo Scientific Chemicals
3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L12958.22
also known as L12958-22
100 g
Catalog number L12958.22
also known as L12958-22
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.40
Save 11.75 (14%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 30-Jun-2026
82.40
Save 11.75 (14%)
Each
Specifications
Beilstein1574570
CAS1455-42-1
Chemical Name or Material3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane
Formula Weight304.39
IUPAC Name2-[3-(1-hydroxy-2-methylpropan-2-yl)-2,4,8,10-tetraoxaspiro[5.5]undecan-9-yl]-2-methylpropan-1-ol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only