(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%
Thermo Scientific Chemicals

(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%

CAS: 29841-69-8 | C14H16N2 | 212.296 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L12968.03
also known as L12968-03
Price (USD)
58.65
Online Exclusive
64.70
Save 6.05 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
58.65
Online Exclusive
64.70
Save 6.05 (9%)
Each
Chemical Identifiers
CAS29841-69-8
IUPAC Name(1S,2S)-1,2-diphenylethane-1,2-bis(aminium)
Molecular FormulaC14H18N2
InChI KeyPONXTPCRRASWKW-KBPBESRZSA-P
SMILES[NH3+][C@H]([C@@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)White to pale cream to cream to yellow
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥96.0%
Melting Point (clear melt)77.0-86.0?C
View more
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air.
RUO – Research Use Only

General References:

  1. Takeshi Ohkuma.; Hirohito Ooka.; Shohei Hashiguchi.; Takao Ikariya.; Ryoji Noyori. Practical Enantioselective Hydrogenation of Aromatic Ketones. J. Am. Chem. Soc.. 1995, 117 (9), 2675-2676.
  2. Michael H. Nantz.; David A. Lee.; Daniel M. Bender.; Azeen H. Roohi. An enantioselective synthesis of vicinal diamines. J. Org. Chem. 1992, 57 (24), 6653-6657.
  3. Chiral auxiliary and solvation agent; see preceding entry.
  4. For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), see: J. Org. Chem ., 57, 6653 (1992).
  5. Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones: J. Am. Chem. Soc., 117, 2675 (1995).