3,5-Dibromo-4-methylaniline, 99%
3,5-Dibromo-4-methylaniline, 99%
3,5-Dibromo-4-methylaniline, 99%
Thermo Scientific Chemicals

3,5-Dibromo-4-methylaniline, 99%

CAS: 13194-73-5 | C7H7Br2N | 264.948 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13101.03
also known as L13101-03
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
67.70
Save 10.05 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 30-Jun-2026
67.70
Save 10.05 (15%)
Each
Chemical Identifiers
CAS13194-73-5
IUPAC Name3,5-dibromo-4-methylaniline
Molecular FormulaC7H7Br2N
InChI KeyAQZDIKCNODUMNY-UHFFFAOYSA-N
SMILESCC1=C(Br)C=C(N)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Pale cream to cream to yellow to brown
Assay (GC)≥98.5%
3,5-Dibromo-4-methylaniline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromo-4-methylaniline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Lisa A.Tobin; Carine Robert; Pratik Nagaria; Saranya Chumsri; William Twaddell; Olga B.Ioffe; George E.Greco; Angela H.Brodie; Alan E.Tomkinson; Feyruz V.Rassool. Targeting abnormal DNA repair in therapy-resistant breast cancers. Mol Cancer Res. 2012, 10,(1), 96-107.
  2. Rene Stangenberg; David Turp; Klaus Mullen. Shape persistent hybrid dendrimers from benzene and triazole via ‘click chemistry’. Tetrahedron. 2014, 70,(19), 3178-3184.