3-Methyl-4-nitroaniline, 95%
3-Methyl-4-nitroaniline, 95%
3-Methyl-4-nitroaniline, 95%
3-Methyl-4-nitroaniline, 95%
Thermo Scientific Chemicals

3-Methyl-4-nitroaniline, 95%

CAS: 611-05-2 | C7H8N2O2 | 152.153 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13112.06
also known as L13112-06
Price (USD)
74.65
Online Exclusive
82.60
Save 7.95 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
74.65
Online Exclusive
82.60
Save 7.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS611-05-2
IUPAC Name3-methyl-4-nitroaniline
Molecular FormulaC7H8N2O2
InChI KeyXPAYEWBTLKOEDA-UHFFFAOYSA-N
SMILESCC1=CC(N)=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pale green
FormCrystals or powder or crystalline powder or flakes
Assay (HPLC)≥98.5%
Melting Point (clear melt)132.0-138.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only