4-Hydroxy-2-(trifluoromethyl)pyrimidine, 97%
4-Hydroxy-2-(trifluoromethyl)pyrimidine, 97%
4-Hydroxy-2-(trifluoromethyl)pyrimidine, 97%
4-Hydroxy-2-(trifluoromethyl)pyrimidine, 97%
Thermo Scientific Chemicals

4-Hydroxy-2-(trifluoromethyl)pyrimidine, 97%

CAS: 1546-80-1 | C5H3F3N2O | 164.087 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13118.06
also known as L13118-06
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1546-80-1
IUPAC Name2-(trifluoromethyl)-3,4-dihydropyrimidin-4-one
Molecular FormulaC5H3F3N2O
InChI KeyPDCVDVCPQWFGAX-UHFFFAOYSA-N
SMILESFC(F)(F)C1=NC=CC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to red
Assay (HPLC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only