6-Heptenoic acid, 96%
6-Heptenoic acid, 96%
6-Heptenoic acid, 96%
6-Heptenoic acid, 96%
Thermo Scientific Chemicals

6-Heptenoic acid, 96%

CAS: 1119-60-4 | C7H12O2 | 128.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13144.03
also known as L13144-03
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1119-60-4
IUPAC Namehept-6-enoic acid
Molecular FormulaC7H12O2
InChI KeyRWNJOXUVHRXHSD-UHFFFAOYSA-N
SMILESOC(=O)CCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (GC)≥95.0%
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
View more
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

General References:

  1. Nisha; Kewal, K.; Vipan, K. Prodigiosin alkaloids: recent advancements in total synthesis and their biological potential. RSC Adv. 2015, 5 (15), 10899-10920.
  2. Wei, S.; Li, Y.; Ma, Q.; Lou, S.; Schreiner, M. Characterization of Tang Dynasty lamp oil remains by using pyrolysis gas chromatography and mass spectrometry. J. Anal. Appl. Pyrolysis 2015, 116, 237-242.