1-Phenyl-1,4-pentanedione, 96%
1-Phenyl-1,4-pentanedione, 96%
1-Phenyl-1,4-pentanedione, 96%
1-Phenyl-1,4-pentanedione, 96%
Thermo Scientific Chemicals

1-Phenyl-1,4-pentanedione, 96%

CAS: 583-05-1 | C11H12O2 | 176.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13151.06
also known as L13151-06
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
109.65
Special offer
Online exclusive
Ends: 30-Jun-2026
129.00
Save 19.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS583-05-1
IUPAC Name1-phenylpentane-1,4-dione
Molecular FormulaC11H12O2
InChI KeyRBLXWIPBPPVLPU-UHFFFAOYSA-N
SMILESCC(=O)CCC(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥95.0%
Identification (FTIR)Conforms
Refractive Index1.5295-1.5360 @20°C
Appearance (Color)Clear colourless to yellow or pale orange
1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Phenyl-1,4-pentanedione is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Matt McLeod; Nicolas Boudreault; Yves Leblanc. Synthetic application of monoprotected hydrazines toward the synthesis of 1-aminopyrroles. J. Org. Chem. 1996, 61,(3), 1180-1183.
  2. Wael Baidossi; Michal Lahav; Jochanan Blum. Hydration of alkynes by a PtCl4-CO catalyst. J. Org. Chem. 1997, 62,(3), 669-672.