Acridine Orange
Acridine Orange
Acridine Orange
Thermo Scientific Chemicals

Acridine Orange

CAS: 10127-02-3 | C17H20Cl3N3Zn | 438.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13159.14
also known as L13159-14
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.80
Save 11.15 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
76.80
Save 11.15 (15%)
Each
Chemical Identifiers
CAS10127-02-3
IUPAC Namezinc(2+) hydrogen N3,N3,N6,N6-tetramethylacridine-3,6-diamine trichloride
Molecular FormulaC17H20Cl3N3Zn
InChI KeyVADJQOXWNSPOQA-UHFFFAOYSA-L
SMILES[H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSuitable for microscopy
Infrared spectrumConforms
Loss on Drying≤10.0% (105°C, 1 hr, US-sourced material)
Loss on Drying≤10.0% (200°C/constant wt, UK-sourced material)
lambda-max (UV-Vis)490-495nm (in water, UK-sourced material)
View more
Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Biological stain.
  2. Selvaggi, R.; Tarpani, L.; Santuari, A.; Giovagnoli, S.; Latterini, L. Silica nanoparticles assisted photodegradation of acridine orange in aqueous suspensions. Appl. Catal., B 2015, 168-169, 363-369.
  3. Zhang, Y.; Pan, J.; Zhang, G.; Zhou, X. Intercalation of herbicide propyzamide into DNA using acridine orange as a fluorescence probe. Sens. Actuators, B 2015, 206, 630-639.