1-Butanol, 99%
1-Butanol, 99%
1-Butanol, 99%
Thermo Scientific Chemicals

1-Butanol, 99%

CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1000 mL
2500 mL
Catalog number L13171.0F
also known as L13171-0F
Price (USD)
82.90
Each
Quantity:
2500 mL
Request bulk or custom format
Price (USD)
82.90
Each
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.3975-1.4005 @ 20?C
FormLiquid
Identification (FTIR)Conforms
1-Butanol is used as a solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as a mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Butanol is used as a solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as a mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

Solubility
Miscible with alcohols, ketones, aldehydes, ethers, aliphatic and aromatic hydrocarbons. Slightly miscible with water.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents, alkali metals and halogens.
RUO – Research Use Only

General References:

  1. Wang, M.; Hu, L.; Fan, L.; Tan, T. Enhanced 1-Butanol Production in Engineered Klebsiella pneumoniaeby NADH Regeneration. Energy Fuels 2015, 29 (3), 1823-1829.
  2. Zhang, X.; Yang, B.; Yuan, W.; Cheng, Z.; Zhang, L.; Li, Y.; Qi, F. Pyrolysis of 2-methyl-1-butanol at low and atmospheric pressures: Mass spectrometry and modeling studies. Proc. Combust. Inst. 2010, 35 (1), 409-417.