Barium manganate, tech. 90%
Barium manganate, tech. 90%
Barium manganate, tech. 90%
Thermo Scientific Chemicals

Barium manganate, tech. 90%

CAS: 7787-35-1 | BaMnO4 | 256.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13172.06
also known as L13172-06
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.00
Save 5.35 (16%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
28.65
Special offer
Online exclusive
Ends: 30-Jun-2026
34.00
Save 5.35 (16%)
Each
Chemical Identifiers
CAS7787-35-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale blue to dark blue or dark green to black
FormPowder or Chunks
Assay41.0-52.2% (MnO4)
Assay (unspecified)41.0-52.2% (MnO4)
Loss on Drying≤1.5% (105°C, 1 hour)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only