FormCrystals or powder or crystalline powder
CommentSpecification differs for U.S. and non-U.S. material where indicated
Optical Rotation139.4 ± 0.1° (C=1 in ethanol, non-U.S. specification)
Water Content (Karl Fischer Titration)≤3%
Appearance (Color)White to off-white
View more