Hexanes, mixed isomers, 98+%
Hexanes, mixed isomers, 98+%
Hexanes, mixed isomers, 98+%
Thermo Scientific Chemicals

Hexanes, mixed isomers, 98+%

CAS: 92112-69-1 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1000 mL
2500 mL
Catalog number L13233.AU
also known as L13233-AU
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.80
Save 7.15 (14%)
Each
Quantity:
1000 mL
Request bulk or custom format
Price (USD)
42.65
Special offer
Online exclusive
Ends: 30-Jun-2026
49.80
Save 7.15 (14%)
Each
Chemical Identifiers
CAS92112-69-1
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3775-1.3815 @ 20?C
Assay (GC)≥98.0% (sum of isomers)
Appearance (Color)Clear colorless
Identification (FTIR)Conforms
FormLiquid
Hexane mixed isomers are used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds, namely butyl lithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexane mixed isomers are used in the laboratory to extract oil and grease contaminants from water and soil for analysis. As a non-polar solvent, it is used in chromatography. It plays an important role in the preparation of glues for shoes, leather products, and roofing. It is also used for reactions that involve very strong bases such as the preparation of Grignard reagents. Also involved in the preparation of organolithiums compounds, namely butyl lithium.

Solubility
Miscible with alcohol, chloroform, acetone and ether. Immiscible with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents. Keep away from sources of ignition.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Herm, Z. R.; Wiers, B. M.; Mason, J. A.; van Baten, J. M.; Hudson, M. R.; Zajdel, P.; Brown, C. M.; Masciocchi, N.; Krishna, R.; Long, J. R. Separation of Hexane Isomers in a Metal-Organic Framework with Triangular Channels. Science 2013, 340 (6135), 960-964.
  2. Titze, T.; Chmelik, C.; Karger, J.; van Baten, J. M.; Krishna, R. Uncommon Synergy between Adsorption and Diffusion of Hexane Isomer Mixtures in MFI Zeolite Induced by Configurational Entropy Effects. J. Phys. Chem. C 2014, 118 (5), 2660-2665.
  3. Deng, Z.; Pera-Titus, M.; Guo, Y.; Giroir-Fendler, A. Molecular Sieving Separation of Hexane Isomers within Nanocomposite (B)-MFI-Alumina Hollow Fiber Membranes: A Modeling Study. Ind. Eng. Chem. Res. 2010, 49 (22), 11697-11707.