Potassium hydride, 30% w/w in mineral oil
Potassium hydride, 30% w/w in mineral oil
Thermo Scientific™

Potassium hydride, 30% w/w in mineral oil

CAS: 7693-26-7 | HK | 40.11 g/mol
Have Questions?
Catalog number L13266.NB
also known as L13266-NB
Price (USD)
338.00
Each
Add to cart
Request bulk or custom format
Price (USD)
338.00
Each
Add to cart
Chemical Identifiers
CAS7693-26-7
IUPAC Namepotassium hydride
Molecular FormulaHK
InChI KeyOCFVSFVLVRNXFJ-UHFFFAOYSA-N
SMILES[H-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)28-32% w/w KH
Appearance (Color)Pale grey or pale brown
FormSuspension in mineral oil
Impurity contentElemental K (as wt% on KH) < 5.0%
Potassium hydride is a precursor in the production of various bases by deprotonation, highly-hindered bases potassium triethylmethoxide and potassium 3-aminopropylamide It serves as a powerful base in organic synthesis of deprotonate carbonyl compounds and amines to form enolates and amides respectively.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.