Sodium peroxide, 95%
Sodium peroxide, 95%
Sodium peroxide, 95%
Thermo Scientific Chemicals

Sodium peroxide, 95%

CAS: 1313-60-6 | Na2O2 | 77.978 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L13306.0B
also known as L13306-0B
Price (USD)
558.00
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
558.00
Each
Chemical Identifiers
CAS1313-60-6
IUPAC Namedisodium dioxidanediide
Molecular FormulaNa2O2
InChI KeyPFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][O-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormGranular solid or beads
Assay (Redox Titration)≥ 94.0% to ≤ 106.0%
Appearance (Color)White to yellow
CommentMay contain a small quantity of discoloured material from the manufacturing process.
Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium peroxide acts as a bleaching agent in laundry and wood pulp. It finds application in the extraction of minerals from various ores. It acts as an oxidizing agent as well as an oxygen source by reacting with carbon dioxide. Furthermore, it is used as a deodorant and an analytical reagent. It is also useful in scuba gear and in submarines.

Solubility
Soluble in acid. Insoluble in alkali.

Notes
Air sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. It reacts with water violently to give sodium hydroxide and hydrogen peroxide. Incompatible with strong reducing agents, organic materials, alcohols and powdered metals.
RUO – Research Use Only

General References:

  1. For use in the direct oxidation of benzaldoximes to acids, see: Synthesis, 807 (1976).
  2. Wu, Y.; Wang, B.; Zhang, Q.; Li, R.; Yu, J. A novel process for high efficiency recovery of rare earth metals from waste phosphors using a sodium peroxide system. RSC Adv. 2014, 4 (16), 7927-7932.
  3. Wei, X. J.; Tian, Z. Q. Simultaneous Determination of Ruthenium and Zinc in Catalysts for Hydrogenation of Benzene to Cyclohexene Using Sodium Peroxide Fusion Sample Digestion and ICP-OES. Adv. Mat. Res. 2014, 1004, 1281-1284.