(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 98+%
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 98+%
(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 98+%
Thermo Scientific Chemicals

(+/-)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 98+%

CAS: 20007-72-1 | C11H14O2 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13326.03
also known as L13326-03
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS20007-72-1
IUPAC Name8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione
Molecular FormulaC11H14O2
InChI KeyDNHDRUMZDHWHKG-UHFFFAOYNA-N
SMILESCC12CCC(=O)C=C1CCCC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point45-52°C
Appearance (Color)White to cream or pale brown
FormPowder or crystalline powder or lumps
Assay (HPLC)≥98.0%
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only