Tri(2-furyl)phosphine, 97%
Tri(2-furyl)phosphine, 97%
Tri(2-furyl)phosphine, 97%
Tri(2-furyl)phosphine, 97%
Thermo Scientific Chemicals

Tri(2-furyl)phosphine, 97%

CAS: 5518-52-5 | C12H9O3P | 232.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L13329.06
also known as L13329-06
Price (USD)
233.65
Online Exclusive
259.00
Save 25.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
233.65
Online Exclusive
259.00
Save 25.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS5518-52-5
IUPAC Nametris(furan-2-yl)phosphane
Molecular FormulaC12H9O3P
InChI KeyDLQYXUGCCKQSRJ-UHFFFAOYSA-N
SMILESO1C=CC=C1P(C1=CC=CO1)C1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or yellow to orange
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
Melting Point57.5-66.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only