(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%
(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%
(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%
Thermo Scientific Chemicals

(S)-(+)-3,4,8,8a-Tetrahydro-8a-methyl-1,6(2H,7H)-naphthalenedione, 99%

CAS: 33878-99-8 | C11H14O2 | 178.231 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
2 g
Catalog number L13334.ME
also known as L13334-ME
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Quantity:
500 mg
Request bulk or custom format
Price (USD)
118.65
Online Exclusive
132.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS33878-99-8
IUPAC Name8a-methyl-1,2,3,4,6,7,8,8a-octahydronaphthalene-1,6-dione
Molecular FormulaC11H14O2
InChI KeyDNHDRUMZDHWHKG-UHFFFAOYNA-N
SMILESCC12CCC(=O)C=C1CCCC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Enantiomeric excess≥98.5%
Optical Rotation+99.4? ± 1.0? (c=1 in benzene)
FormCrystals or powder or crystalline powder or granules
Assay (GC)≥98.5%
Melting Point (clear melt)47.0-53.0?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only