1-Boc-piperazine, 99%
1-Boc-piperazine, 99%
1-Boc-piperazine, 99%
Thermo Scientific Chemicals

1-Boc-piperazine, 99%

CAS: 57260-71-6 | C9H19N2O2 | 187.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L13363.03
also known as L13363-03
Price (USD)
33.65
Online Exclusive
37.00
Save 3.35 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
33.65
Online Exclusive
37.00
Save 3.35 (9%)
Each
Chemical Identifiers
CAS57260-71-6
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or crystalline powder or lumpy powder or fused solid
Assay (GC)≥98.5%
Appearance (Color)White to cream to yellow
Solution Test5% in ethanol: clear & colourless to yellow
1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperazine is useful for the preparation of (m-phenoxy)phenyl substituted piperazine derivatives. It is also used in the synthesis of indazole DNA gyrase inhibitors. Further, it plays an important role in the preparation of alfa,beta-poly(2-oxazoline) lipopolymers through living cationic ring opening polymerization. It is also in involved in Buchwald-Hartwig coupling reactions with aryl halides.

Solubility
Soluble in ethyl acetate, methanol and water.

Notes
Hygroscopic. Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Pd-catalyzed C-N coupling with a variety of bromoarenes, in the presence of NaO-t-Bu and Tri(o-tolyl) phosphine, A12093, provides a route to arylpiperazines of potential pharmacological interest: Tetrahedron Lett., 39, 2219 (1998).
  2. Ba-Salem, A. O.; Shaikh, M. N.; Ullah, N.; Faiz, M. Synthesis and magnetic relaxation properties of new Gd(III) complexes derived from DTPA-bis(amide) conjugates of arylpiperazinyl amines. Inorg. Chem. Commun. 2015, 56, 5-7.
  3. Nemeckova-Herova, D.; Pazdera, P. A Simplified Protocol for Routine Chemoselective Syntheses of Piperazines Substituted in the 1-Position by an Electron Withdrawing Group. Curr. Org. Synth. 2015, 12 (2), 173-179.