2-Fluoro-5-methylaniline, 98%
2-Fluoro-5-methylaniline, 98%
2-Fluoro-5-methylaniline, 98%
Thermo Scientific Chemicals

2-Fluoro-5-methylaniline, 98%

CAS: 452-84-6 | C7H8FN | 125.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13411.06
also known as L13411-06
Price (USD)
25.40
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
25.40
Each
Chemical Identifiers
CAS452-84-6
IUPAC Name2-fluoro-5-methylaniline
Molecular FormulaC7H8FN
InChI KeyQZUXMXZNVAJNSE-UHFFFAOYSA-N
SMILESCC1=CC=C(F)C(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow to yellow to red or purple to dark pink to pink
Refractive Index1.5310-1.5350 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only