N-(n-Butyl)ethylenediamine, 97%
N-(n-Butyl)ethylenediamine, 97%
N-(n-Butyl)ethylenediamine, 97%
N-(n-Butyl)ethylenediamine, 97%
Thermo Scientific Chemicals

N-(n-Butyl)ethylenediamine, 97%

CAS: 19522-69-1 | C6H16N2 | 116.208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13437.06
also known as L13437-06
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS19522-69-1
IUPAC Name(2-aminoethyl)(butyl)amine
Molecular FormulaC6H16N2
InChI KeyDFPGBRPWDZFIPP-UHFFFAOYSA-N
SMILESCCCCNCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4395-1.4445 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only