4-Methyl-3-nitrobenzaldehyde, 98%
4-Methyl-3-nitrobenzaldehyde, 98%
4-Methyl-3-nitrobenzaldehyde, 98%
Thermo Scientific Chemicals

4-Methyl-3-nitrobenzaldehyde, 98%

CAS: 31680-07-6 | C8H7NO3 | 165.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L13454.03
also known as L13454-03
Price (USD)
30.65
Online Exclusive
34.30
Save 3.65 (11%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
34.30
Save 3.65 (11%)
Each
Chemical Identifiers
CAS31680-07-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
Free acid (titration)≤1.5%
4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-3-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It is sensitive to air.
RUO – Research Use Only

General References:

  1. Guo, Z-R.; H-B. Li.; Fang Li. 4-Methyl-3-nitro­benzaldehyde. Acta Cryst. 2010, 66 2420.
  2. Yadav, B S.; Tyagi, S K.; Seema. Study of vibrational spectra of 4-methyl-3-nitrobenzaldehyde. IJPAP. 2006, 44 (9), 644-648.