2-Chloroaniline hydrochloride, 97%
2-Chloroaniline hydrochloride, 97%
2-Chloroaniline hydrochloride, 97%
Thermo Scientific Chemicals

2-Chloroaniline hydrochloride, 97%

CAS: 137-04-2 | C6H7Cl2N | 164.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L13483.14
also known as L13483-14
Price (USD)
40.65
Online Exclusive
44.70
Save 4.05 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
40.65
Online Exclusive
44.70
Save 4.05 (9%)
Each
Chemical Identifiers
CAS137-04-2
IUPAC Namehydrogen 2-chloroaniline chloride
Molecular FormulaC6H7Cl2N
InChI KeyDRGIDRZFKRLQTE-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (GC)≥96.0%
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. E Hü.; G Bereket.; Y Şahin. Corrosion inhibition of stainless steel by polyaniline, poly (2-chloroaniline), and poly (aniline-co-2-chloroaniline) in HCl. Progress in organic coatings. 2006, 57,(2), 149-158.
  2. KF Khaled.; N Hackerman. Investigation of the inhibitive effect of ortho-substituted anilines on corrosion of iron in 1 M HClsolutions. Electrochimica Acta. 2003, 48,(19), 2715-2723.