2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol
2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol
2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol
Thermo Scientific Chemicals

2-(Dimethylamino)ethyl acrylate, 98%, stab. with ca 0.1% 4-methoxyphenol

CAS: 2439-35-2 | C7H13NO2 | 143.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Catalog number L13506.AE
also known as L13506-AE
Price (USD)
64.65
Online Exclusive
71.30
Save 6.65 (9%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
64.65
Online Exclusive
71.30
Save 6.65 (9%)
Each
Chemical Identifiers
CAS2439-35-2
IUPAC Name2-(dimethylamino)ethyl prop-2-enoate
Molecular FormulaC7H13NO2
InChI KeyDPBJAVGHACCNRL-UHFFFAOYSA-N
SMILESCN(C)CCOC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pale brown
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4360-1.4400 @ 20°C
Solution Test(10% v/v in methanol) : Clear
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)ethyl acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, as a catalytic agent, as a petrochemical additive and agrochemicals.

Solubility
Soluble in water (1.000 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Sensitive to heat, light and moisture.
RUO – Research Use Only

General References:

  1. Kejian Bian.; Michael F. Cunningham. Nitroxide-mediated radical polymerization of 2-(dimethylamino)ethyl acrylate and its sequential block copolymerization with styrene and N-butyl acrylate. Journal of Polymer Science Part A: Polymer Chemistry. 2006, 44 (1), 414-426.
  2. M.S. Aziz.; Y.A. Aggourb. Electrical conduction of polymer complexes of 2-(dimethylamino)ethyl acrylate with cobalt chloride. Polymer Testing. 1999, 18 (7), 511-521.