2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
Thermo Scientific Chemicals

2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol

CAS: 2867-47-2 | C8H15NO2 | 157.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L13507.AE
also known as L13507-AE
Price (USD)
35.65
Online Exclusive
39.90
Save 4.25 (11%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
35.65
Online Exclusive
39.90
Save 4.25 (11%)
Each
Chemical Identifiers
CAS2867-47-2
IUPAC Name2-(dimethylamino)ethyl 2-methylprop-2-enoate
Molecular FormulaC8H15NO2
InChI KeyJKNCOURZONDCGV-UHFFFAOYSA-N
SMILESCN(C)CCOC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.4375-1.4425 @ 20?C
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.

Solubility
Soluble in water. (1.000 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture and light. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. J Y Cherng.; P van de Wetering.; H Talsma.; D J Crommelin.; W E Hennink. Freeze-drying of poly((2-dimethylamino)ethyl methacrylate)-based gene delivery systems. Pharmaceutical Research. 1997, 14 (12), 1838-1841.
  2. Sang Beom Lee.; Alan J Russell.; Krzysztof Matyjaszewski. ATRP synthesis of amphiphilic random, gradient, and block copolymers of 2-(dimethylamino)ethyl methacrylate and n-butyl methacrylate in aqueous media. Biomacromolecules. 2003, 4 (5), 1386-1393.