3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals
3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals
3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals
3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Acetyl-2-methyl-5-phenylthiophene, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L13554.06
also known as L13554-06
5 g
Catalog number L13554.06
also known as L13554-06
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
165.65
Online Exclusive
184.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS40932-63-6
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Cream to pale brown
FormCrystals or powder or crystalline powder
Melting Point (clear melt)65.5-71.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only