5-Bromo-3-pyridineacetic acid, 98+%
5-Bromo-3-pyridineacetic acid, 98+%
5-Bromo-3-pyridineacetic acid, 98+%
Thermo Scientific Chemicals

5-Bromo-3-pyridineacetic acid, 98+%

CAS: 39891-12-8 | C7H6BrNO2 | 216.034 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13579.03
also known as L13579-03
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
104.65
Online Exclusive
116.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS39891-12-8
IUPAC Name2-(5-bromopyridin-3-yl)acetic acid
Molecular FormulaC7H6BrNO2
InChI KeyUETIDNDXXGCJCE-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC(Br)=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)Cream to orange to brown or pink
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-3-pyridineacetic acid acts as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff field.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Yao, X.; Diego, P.; Ramos, A. A.; Shi, Y. Averagine-Scaling Analysis and Fragment Ion Mass Defect Labeling in Peptide Mass Spectrometry. Anal. Chem. 2008, 80 (19), 7383-7391.
  2. Zhang, J.; Huitema, C.; Niu, C.; Yin, J.; James, M. N. G.; Eltis, L. D.; Vederas, J. C. Aryl methylene ketones and fluorinated methylene ketones as reversible inhibitors for severe acute respiratory syndrome (SARS) 3C-like proteinase. Bioorg. Chem. 2008, 36 (5), 229-240.