1-Bromo-4-(ethylthio)benzene, 97%
1-Bromo-4-(ethylthio)benzene, 97%
1-Bromo-4-(ethylthio)benzene, 97%
1-Bromo-4-(ethylthio)benzene, 97%
Thermo Scientific Chemicals

1-Bromo-4-(ethylthio)benzene, 97%

CAS: 30506-30-0 | C8H9BrS | 217.124 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L13718.14
also known as L13718-14
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.00
Save 13.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
89.00
Save 13.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS30506-30-0
IUPAC Name1-bromo-4-(ethylsulfanyl)benzene
Molecular FormulaC8H9BrS
InChI KeyCTFHVAKXUMMGOC-UHFFFAOYSA-N
SMILESCCSC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.6045-1.6095 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only