5-Fluoro-2-methylbenzoyl chloride, 98%
5-Fluoro-2-methylbenzoyl chloride, 98%
5-Fluoro-2-methylbenzoyl chloride, 98%
Thermo Scientific Chemicals

5-Fluoro-2-methylbenzoyl chloride, 98%

CAS: 21900-39-0 | C8H6ClFO | 172.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13750.03
also known as L13750-03
Price (USD)
28.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
28.10
Each
Chemical Identifiers
CAS21900-39-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5305-1.5345 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only