2-Acetylbenzo[b]furan, 99%
2-Acetylbenzo[b]furan, 99%
2-Acetylbenzo[b]furan, 99%
Thermo Scientific Chemicals

2-Acetylbenzo[b]furan, 99%

CAS: 1646-26-0 | C10H8O2 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13771.14
also known as L13771-14
Price (USD)
192.65
Online Exclusive
214.00
Save 21.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
192.65
Online Exclusive
214.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS1646-26-0
IUPAC Name1-(1-benzofuran-2-yl)ethan-1-one
Molecular FormulaC10H8O2
InChI KeyYUTFQTAITWWGFH-UHFFFAOYSA-N
SMILESCC(=O)C1=CC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)68.0-74.0?C
Appearance (Color)White to cream to yellow to brown to gray
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥98.5%
2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. J Kossakowski.; K Ostrowska.; E Hejchman.; I Wolska - Il. Synthesis and structural characterization of derivatives of 2-and 3-benzo [b] furan carboxylic acids with potential cytotoxic activity. Farmaco. 2005, 60,(6-7), 519-527.
  2. K Ando.; Y Kawamura.; Y Akai.; J Kunitomo. Preparation of 2-, 3-, 4-and 7-(2-alkylcarbamoyl-1-alkylvinyl) benzo [b] furans and their BLT 1 and/or BLT 2 inhibitory activities. Organic & Biomolecular Chemistry. 2005, 6,(2), 296-307.